Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Avpr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011924.2 | NP_036054.1 | 1 - 2 | 1557 | 69 | |
GenBank mRNA | AK033416.1 | - | 1 - 2 | 591 | 69 | |
AK050523.1 | - | 1 - 2 | 639 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mus musculus Probe spans exons |