Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204239.1 | NP_001191168.1 | 3 - 4 | 663 | 112 | |
NM_019985.3 | NP_064369.1 | 4 - 5 | 759 | 112 | ||
GenBank mRNA | AF201457.1 | - | 4 - 5 | 470 | 112 | |
AK007925.1 | - | 4 - 5 | 112 | |||
BC064054.1 | - | 4 - 5 | 759 | 574821 | 112 | |
EF070191.1 | - | 3 - 4 | 353 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |