Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023456.2 | NP_075945.1 | 2 - 3 | 259 | 107 | |
GenBank mRNA | AF273768.1 | - | 1 - 2 | 186 | 107 | |
AF286198.1 | - | 1 - 2 | 186 | 107 | ||
AF286199.1 | - | 1 - 2 | 186 | 107 | ||
AK002982.1 | - | 2 - 3 | 272 | 107 | ||
BC043012.1 | - | 2 - 3 | 259 | 5683102 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |