Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284412.1 | NP_001271341.1 | 2 - 3 | 252 | 122 | |
NM_001284414.1 | NP_001271343.1 | 2 - 3 | 252 | 122 | ||
NM_026776.4 | NP_081052.2 | 2 - 3 | 252 | 122 | ||
GenBank mRNA | AK002500.1 | - | 2 - 3 | 214 | 122 | |
AK003715.2 | - | 2 - 3 | 224 | 122 | ||
AK003877.1 | - | 2 - 3 | 210 | 122 | ||
AK082933.1 | - | 2 - 3 | 231 | 122 | ||
AK145811.1 | - | 2 - 3 | 223 | 122 | ||
AK150822.1 | - | 2 - 3 | 234 | 122 | ||
AK162230.1 | - | 2 - 3 | 225 | 122 | ||
AK166008.1 | - | 2 - 3 | 231 | 122 | ||
BC029181.1 | - | 2 - 3 | 216 | 4240500 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |