Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021354.1 | NP_067329.1 | 11 - 12 | 1024 | 96 | |
GenBank mRNA | AJ243590.1 | - | 11 - 12 | 1024 | 96 | |
AK012988.1 | - | 11 - 12 | 992 | 96 | ||
BC082564.1 | - | 11 - 12 | 980 | 30551418 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |