Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sarnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025364.2 | NP_079640.1 | 6 - 7 | 542 | 92 | |
GenBank mRNA | AK003453.1 | - | 6 - 7 | 394 | 92 | |
AK018773.1 | - | 6 - 7 | 405 | 92 | ||
AK088266.1 | - | 6 - 7 | 391 | 92 | ||
AK146248.1 | - | 6 - 7 | 420 | 92 | ||
BC027510.1 | - | 6 - 7 | 381 | 1382354 | 92 | |
BC094393.1 | - | 6 - 7 | 378 | 92 | ||
BC157909.1 | - | 6 - 7 | 406 | 9088111 | 92 | |
BC158023.1 | - | 6 - 7 | 406 | 9007329 | 92 | |
BC172063.1 | - | 6 - 7 | 403 | 9054349 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |