Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sarnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025364.2 | NP_079640.1 | 8 - 9 | 591 | 109 | |
GenBank mRNA | AK003453.1 | - | 8 - 9 | 443 | 109 | |
AK018773.1 | - | 8 - 9 | 454 | 109 | ||
AK088266.1 | - | 8 - 9 | 440 | 109 | ||
AK146248.1 | - | 8 - 9 | 469 | 109 | ||
BC027510.1 | - | 8 - 9 | 430 | 1382354 | 109 | |
BC157909.1 | - | 8 - 9 | 455 | 9088111 | 109 | |
BC158023.1 | - | 8 - 9 | 455 | 9007329 | 109 | |
BC172063.1 | - | 8 - 9 | 452 | 9054349 | 109 | |
BF148013.1 | - | 3 - 4 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |