Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ero1lb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026184.2 | NP_080460.2 | 14 - 15 | 1409 | 105 | |
GenBank mRNA | AF489856.1 | - | 14 - 15 | 1215 | 105 | |
AK004981.1 | - | 1386 | 105 | |||
AK006886.1 | - | 14 - 15 | 1269 | 105 | ||
AK011964.1 | - | 282 | 105 | |||
AK041951.1 | - | 14 - 15 | 1385 | 105 | ||
AK160664.1 | - | 14 - 15 | 1425 | 105 | ||
BC058721.1 | - | 14 - 15 | 1409 | 5705507 | 105 | |
BC111900.1 | - | 14 - 15 | 1379 | 40062137 | 105 | |
BC112426.1 | - | 14 - 15 | 1379 | 40062136 | 105 | |
BC145888.1 | - | 14 - 15 | 1360 | 40131191 | 105 | |
BC145890.1 | - | 14 - 15 | 1360 | 40131193 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |