Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsix | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_002844.2 | 3 - 4 | 622 | 74 | |||
GenBank mRNA | AF138745.1 | - | LIT1604 Tsix |
3 - 4 | 622 | 74 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | testis specific X-linked gene | Tsx | AV207184 | -23.6kb | Sense | Chr. X - 103409101 - 103424588 on NCBI Build GRCm38 |
Downstream |
Jpx transcript, Xist activator (non-protein coding) |
Jpx |
2010000I03Rik, 2510040I06Rik, Enox |
8.6kb | Sense | Chr. X - 103493576 - 103506425 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |