Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 10714 | Gm10714 | ENSMUSG00000074555 | -82.5kb | Sense | Chr. 2 - 174125038 - 174183918 on NCBI Build GRCm38 |
Downstream | microRNA 296 | Mir296 |
Mirn296, mir-296, mmu-mir-296 |
11.0 | Sense | Chr. 2 - 174267047 - 174267125 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |