Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meg3 | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027652.1 | 5 - 6 | 1173 | 67 | |||
GenBank mRNA | Y13832.1 | - | LIT3453 Gtl2 |
5 - 6 | 1164 | 67 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 1906-1 | Mir1906-1 |
Mir1906, Mirn1906, mmu-mir-1906, mmu-mir-1906-1 |
-1.8kb | Sense | Chr. 12 - 109544541 - 109544620 on NCBI Build GRCm38 |
Downstream | microRNA 770 | Mir770 |
Mirn770, mir-770, mmu-mir-770 |
14.3kb | Sense | Chr. 12 - 109563692 - 109563785 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |