Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn2r29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003555.1 | 1 - 2 | 194 | 107 | ||
GenBank mRNA | AK046541.1 | - | 1 - 2 | 119 | 107 | |
AK084962.1 | - | 1 - 2 | 119 | 107 | ||
BC096375.1 | - | 1 - 2 | 120 | 107 | ||
BY089007.1 | - | 1 - 2 | 195 | 107 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | zinc finger protein 772 | Zfp772 | -59.3kb | Sense | Chr. 7 - 7202122 - 7209998 on NCBI Build GRCm38 | |
Downstream | chloride channel 4-2 | Clcn4-2 |
Clc4-2, Clcn4 |
4.0kb | Sense | Chr. 7 - 7282309 - 7300851 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |