Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134101.2 | NP_598862.1 | 4 - 5 | 535 | 85 | |
NR_027485.1 | 4 - 5 | 627 | 85 | |||
GenBank mRNA | AK146482.1 | - | 4 - 5 | 484 | 85 | |
AK150374.1 | - | 4 - 5 | 507 | 85 | ||
AK151588.1 | - | 4 - 5 | 507 | 85 | ||
AK152007.1 | - | 4 - 5 | 507 | 85 | ||
AK152970.1 | - | 4 - 5 | 504 | 85 | ||
AK159098.1 | - | 4 - 5 | 487 | 85 | ||
AK159636.1 | - | 4 - 5 | 484 | 85 | ||
AK159756.1 | - | 4 - 5 | 507 | 85 | ||
AK159815.1 | - | 4 - 5 | 503 | 85 | ||
AK166816.1 | - | 4 - 5 | 487 | 85 | ||
AK167326.1 | - | 4 - 5 | 487 | 85 | ||
AK167774.1 | - | 4 - 5 | 484 | 85 | ||
AK167786.1 | - | 4 - 5 | 485 | 85 | ||
AK167909.1 | - | 4 - 5 | 495 | 85 | ||
AK167995.1 | - | 4 - 5 | 503 | 85 | ||
AK168131.1 | - | 4 - 5 | 494 | 85 | ||
AK168398.1 | - | 4 - 5 | 487 | 85 | ||
AK168543.1 | - | 4 - 5 | 484 | 85 | ||
AK169867.1 | - | 4 - 5 | 487 | 85 | ||
AK198780.1 | - | 1 - 2 | 62 | 85 | ||
BC021508.1 | - | 4 - 5 | 504 | 5347139 | 85 | |
BC024058.1 | - | 4 - 5 | 477 | 85 | ||
BC024830.1 | - | 4 - 5 | 475 | 5358424 | 85 | |
BC027388.1 | - | 4 - 5 | 499 | 4952935 | 85 | |
BC027767.1 | - | 4 - 5 | 503 | 5347281 | 85 | |
BC027822.1 | - | 4 - 5 | 486 | 5321505 | 85 | |
BC028851.1 | - | 4 - 5 | 475 | 3660169 | 85 | |
BC031128.1 | - | 4 - 5 | 504 | 4982313 | 85 | |
BC033325.1 | - | 4 - 5 | 564 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |