Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159364.1 | NP_001152836.1 | 3 - 4 | 395 | 72 | |
NM_001159365.1 | NP_001152837.1 | 3 - 4 | 395 | 72 | ||
NM_001159366.1 | NP_001152838.1 | 3 - 4 | 395 | 72 | ||
NM_028815.4 | NP_083091.1 | 3 - 4 | 395 | 72 | ||
GenBank mRNA | AK012971.2 | - | 3 - 4 | 380 | 72 | |
AK040079.1 | - | 3 - 4 | 402 | 72 | ||
AK046486.1 | - | 3 - 4 | 944 | 72 | ||
AK079884.1 | - | 3 - 4 | 363 | 72 | ||
AK080249.1 | - | 3 - 4 | 381 | 72 | ||
AK154885.1 | - | 3 - 4 | 374 | 72 | ||
BC065773.1 | - | 3 - 4 | 344 | 72 | ||
BC067049.1 | - | 3 - 4 | 395 | 30363673 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |