Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mprip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012027.2 | NP_036157.2 | 13 - 14 | 1801 | 101 | |
NM_201245.3 | NP_957697.1 | 13 - 14 | 1801 | 101 | ||
GenBank mRNA | AK035435.1 | - | 7 - 8 | 1091 | 101 | |
AK044512.1 | - | 12 - 13 | 1443 | 101 | ||
AK154807.1 | - | 14 - 15 | 1808 | 101 | ||
AK155630.1 | - | 13 - 14 | 1582 | 101 | ||
AK158296.1 | - | 13 - 14 | 1790 | 101 | ||
AK162530.1 | - | 11 - 12 | 1147 | 101 | ||
AK170849.1 | - | 13 - 14 | 1785 | 101 | ||
AK171089.1 | - | 13 - 14 | 1790 | 101 | ||
BC039932.1 | - | 12 - 13 | 1325 | 101 | ||
BC049803.1 | - | 13 - 14 | 1327 | 101 | ||
BC063067.1 | - | 13 - 14 | 1764 | 5721396 | 101 | |
U73200.1 | - | 13 - 14 | 1661 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |