Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015737.4 | NP_056552.1 | 1 | 1947 | 87 | |
GenBank mRNA | AK033494.1 | - | 1 | 1912 | 87 | |
AK148036.1 | - | 1 | 1915 | 87 | ||
AK153253.1 | - | 1 | 1925 | 87 | ||
BC057882.1 | - | 1 | 1938 | 5350623 | 87 | |
U73819.1 | - | 1 | 1667 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |