Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100028027.mm.1 |
NM_001034097.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028027.mm.1 |
NM_001034097.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028027.mm.1 |
NM_001159503.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028027.mm.1 |
NM_001159503.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028027.mm.1 |
NM_001159505.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028027.mm.1 |
NM_001159505.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028027.mm.1 |
NM_023517.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028027.mm.1 |
NM_023517.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028027.mm.1 |
TC1100003059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028031.mm.1 |
NM_001159505.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028031.mm.1 |
NM_001159505.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028031.mm.1 |
NM_023517.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028031.mm.1 |
NM_023517.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028031.mm.1 |
TC1100003059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028025.mm.1 |
NM_001034097.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028025.mm.1 |
NM_001034097.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028025.mm.1 |
NM_001159503.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028025.mm.1 |
NM_001159503.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028025.mm.1 |
NM_001159505.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028025.mm.1 |
NM_001159505.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028025.mm.1 |
NM_023517.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028025.mm.1 |
NM_023517.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028025.mm.1 |
TC1100003059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028030.mm.1 |
NM_001159505.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028030.mm.1 |
NM_001159505.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028030.mm.1 |
NM_023517.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028030.mm.1 |
NM_023517.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028030.mm.1 |
TC1100003059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028028.mm.1 |
NM_001034097.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028028.mm.1 |
NM_001034097.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028028.mm.1 |
NM_001159503.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028028.mm.1 |
NM_001159503.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028028.mm.1 |
NM_001159505.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028028.mm.1 |
NM_001159505.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028028.mm.1 |
NM_023517.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028028.mm.1 |
NM_023517.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028028.mm.1 |
TC1100003059.mm.1 |
MTA TC |