Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK021139.1 | - | 2 - 3 | 208 | 69 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ZXD family zinc finger C | Zxdc |
A930012H20, B930086F11Rik, BC003332 |
-17.7kb | Anti-sense | Chr. 6 - 90369483 - 90403490 on NCBI Build GRCm38 |
Downstream | Kruppel-like factor 15 | Klf15 |
1810013I09Rik, AV048136, AW494632, CKLF, KKLF |
35.1kb | Anti-sense | Chr. 6 - 90462626 - 90479170 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |