Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Six3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011381.4 | NP_035511.2 | 2 | 105 | ||
GenBank mRNA | AK021385.1 | - | 1 | 149 | 106 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
SIX homeobox 3, opposite strand 1 |
Six3os1 |
D17Mgi26, E130112H22Rik, Rncr1, Six3os |
-5.9kb | Anti-sense | Chr. 17 - 85601932 - 85618396 on NCBI Build GRCm38 |
Downstream | sine oculis-related homeobox 2 | Six2 | 37.5kb | Anti-sense | Chr. 17 - 85662735 - 85688254 on NCBI Build GRCm38 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon |