Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prps1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037746.3 | NP_001032835.2 | 2 | 2922 | 98 | |
GenBank mRNA | AK032842.1 | - | 1 | 1250 | 98 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
solute carrier family 25 (mitochondrial oxodicarboxylate carrier), member 21 |
Slc25a21 |
9930033G19Rik, A630030I10, BB158148 |
-42.4kb | Anti-sense | Chr. 12 - 56712634 - 57197472 on NCBI Build GRCm38 |
Downstream | predicted gene 16246 | Gm16246 | OTTMUSG00000032628 | 198.6kb | Anti-sense | Chr. 12 - 57440360 - 57451337 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon |