Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin28b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK044904.1 | - | 1 | 3348 | 72 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | blood vessel epicardial substance | Bves |
Pop1, Popdc1, mBVES |
-82.5kb | Anti-sense | Chr. 10 - 45335762 - 45372484 on NCBI Build GRCm38 |
Downstream |
HECT domain and ankyrin repeat containing, E3 ubiquitin protein ligase 1 |
Hace1 |
1700042J16Rik, A730034A22Rik, BC025474 |
119.2kb | Anti-sense | Chr. 10 - 45577571 - 45712345 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |