Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnq1ot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_001461.5 | 1 | 1862 | 69 | ||
GenBank mRNA | AK047831.1 | - | 1 | 1855 | 69 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
transient receptor potential cation channel, subfamily M, member 5 |
Trpm5 |
9430099A16Rik, LTrpC-5, Ltrpc5, Mtr1 |
-199.4kb | Sense | Chr. 7 - 143071529 - 143094642 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 4933417O13 gene | 4933417O13Rik | 26.2kb | Sense | Chr. 7 - 143322772 - 143330650 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |