Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK080409.1 | - | 3448 | 67 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
cysteine dioxygenase 1, cytosolic |
Cdo1 |
1300002L19Rik, Cdo, D18Ucla3 |
-1.6kb | Sense | Chr. 18 - 46713205 - 46728542 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |