Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vangl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK082938.1 | - | 1047 | 81 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | SLAM family member 6 | Slamf6 |
KAL1, KAL1b, Ly108, NTB-A, NTBA, SF2000 |
-47.8kb | Anti-sense | Chr. 1 - 171917199 - 171953170 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon |