Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK131677.1 | - | 1 | 553 | 78 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 1700016G14 gene | 1700016G14Rik | -15.0kb | Sense | Chr. 13 - 24733875 - 24746592 on NCBI Build GRCm38 | |
Downstream | cDNA sequence BC005537 | BC005537 |
8030460C05Rik, C78201 |
39.5kb | Sense | Chr. 13 - 24801657 - 24812899 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |