Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK138302.1 | - | 5352 | 97 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome |
---|
Set Membership: |
Amplicon length between 86 and 100 Mus musculus |