Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK011205.1 | - | 1 | 357 | 101 | |
AK143579.1 | - | 1 | 1250 | 101 | ||
AK149041.1 | - | 1 | 843 | 101 | ||
AK169352.1 | - | 1 | 1256 | 101 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | integrin beta 6 | Itgb6 |
2210409C20Rik, 4831415H04Rik |
-27.9kb | Sense | Chr. 2 - 60598292 - 60722603 on NCBI Build GRCm38 |
Downstream | predicted gene 13582 | Gm13582 |
ENSMUSG00000075325, OTTMUSG00000012844 |
213.4kb | Sense | Chr. 2 - 60964376 - 60975151 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |