Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctnnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF140220.1 | - | 1 | 37 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Inventoried Both primers and probe map within a single exon |