Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK042714.1 | - | 1 - 2 | 130 | 79 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
serine (or cysteine) peptidase inhibitor, clade I, member 2 |
Serpini2 | Spi14 | -209.3kb | Sense | Chr. 3 - 75242357 - 75270078 on NCBI Build GRCm38 |
Downstream | programmed cell death 10 | Pdcd10 |
2410003B13Rik, Ccm3, Tfa15, Tfar15 |
34.3kb | Sense | Chr. 3 - 75516490 - 75556852 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons |