Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | activating transcription factor 6 | Atf6 |
9130025P16Rik, 9630036G24, AA789574, Atf6alpha, ESTM49 |
-876.0 | Anti-sense | Chr. 1 - 170704457 - 170867771 on NCBI Build GRCm38 |
Downstream | Fc receptor-like B | Fcrlb |
FREB-2, FREB2, FcRL2, Fcrlm2, Fcry, mFCRL2 |
26.2kb | Anti-sense | Chr. 1 - 170906852 - 170913860 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |