Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | KCNQ1 overlapping transcript 1 | Kcnq1ot1 |
Kvlqt1-as, Lit1, Tssc8 |
-4.4kb | Anti-sense | Chr. 7 - 143213111 - 143296547 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 4933417O13 gene | 4933417O13Rik | 21.2kb | Anti-sense | Chr. 7 - 143322772 - 143330650 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |