Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | group specific component | Gc |
DBP, VDB |
-66.7kb | Anti-sense | Chr. 5 - 89417511 - 89457898 on NCBI Build GRCm38 |
Downstream | neuropeptide FF receptor 2 | Npffr2 |
Gpr74, HG31, NPFF2 |
1.6kb | Sense | Chr. 5 - 89526577 - 89583914 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |