Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm35439 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK137696.1 | - | 1 - 2 | 154 | 95 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | late cornified envelope-like proline-rich 1 | Lelp1 | 1700012F11Rik | -27.5kb | Anti-sense | Chr. 3 - 92134994 - 92142754 on NCBI Build GRCm38 |
Downstream | small proline-rich protein 2A1 | Sprr2a1 |
Sprr2a, Sprr2a3 |
44.5kb | Sense | Chr. 3 - 92215835 - 92222491 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |