Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK049074.1 | - | 2 | 1255 | 80 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 8106 | Mir8106 |
Gm27594, mmu-mir-8106 |
-1.9kb | Sense | Chr. 9 - 122273890 - 122274028 on NCBI Build GRCm38 |
Downstream | abhydrolase domain containing 5 | Abhd5 |
1300003D03Rik, 2010002J10Rik, CDS, CGI-58, IECN5, NCIE2 |
73.4kb | Anti-sense | Chr. 9 - 122351539 - 122381524 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |