Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | mannose-binding lectin (protein A) 1 | Mbl1 |
MBL-A, MBP-A, S-MBP |
-8.2kb | Anti-sense | Chr. 14 - 41151456 - 41159596 on NCBI Build GRCm38 |
Downstream | surfactant associated protein D | Sftpd |
AI573415, SP-D, Sftp4 |
3.8kb | Sense | Chr. 14 - 41172212 - 41185198 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |