Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnase13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011687.2 | NP_001011687.1 | 1 | 256 | 69 | |
GenBank mRNA | BC132614.1 | - | 1 | 256 | 40130891 | 69 |
BC132616.1 | - | 1 | 256 | 40130893 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |