Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012638.1 | NP_001012656.1 | 3 - 4 | 412 | 90 | |
GenBank mRNA | AK032683.1 | - | 3 - 4 | 392 | 90 | |
AK052768.1 | - | 403 | 90 | |||
AK145959.1 | - | 3 - 4 | 397 | 90 | ||
AK160599.1 | - | 3 - 4 | 372 | 90 | ||
AK170606.1 | - | 2 - 3 | 281 | 90 | ||
AK171458.1 | - | 3 - 4 | 403 | 90 | ||
BC138694.1 | - | 3 - 4 | 377 | 8861716 | 90 | |
BC138695.1 | - | 3 - 4 | 377 | 8861717 | 90 | |
BC145384.1 | - | 3 - 4 | 377 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |