Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008506.2 | NP_001008506.2 | 11 - 12 | 1635 | 79 | |
NM_001160107.1 | NP_001153579.1 | 10 - 11 | 1560 | 79 | ||
NM_001160108.1 | NP_001153580.1 | 3 - 4 | 829 | 79 | ||
NM_029334.2 | NP_083610.2 | 13 - 14 | 2028 | 79 | ||
NR_027648.1 | 9 - 10 | 1445 | 79 | |||
GenBank mRNA | AK006009.1 | - | 3 - 4 | 793 | 79 | |
AK048046.1 | - | 13 - 14 | 1994 | 79 | ||
AK050226.1 | - | 13 - 14 | 2009 | 79 | ||
AK076066.1 | - | 9 - 10 | 1441 | 79 | ||
AK160937.1 | - | 10 - 11 | 1546 | 79 | ||
BC024824.1 | - | 10 - 11 | 1530 | 5355748 | 79 | |
BC024856.1 | - | 6 - 7 | 650 | 79 | ||
BC026610.1 | - | 7 - 8 | 1100 | 79 | ||
BC034760.1 | - | 13 - 14 | 1961 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |