Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008506.2 | NP_001008506.2 | 13 - 14 | 1851 | 83 | |
NM_001160107.1 | NP_001153579.1 | 12 - 13 | 1776 | 83 | ||
NM_001160108.1 | NP_001153580.1 | 5 - 6 | 1045 | 83 | ||
NM_029334.2 | NP_083610.2 | 15 - 16 | 2244 | 83 | ||
NR_027648.1 | 11 - 12 | 1649 | 83 | |||
GenBank mRNA | AK006009.1 | - | 5 - 6 | 1009 | 83 | |
AK048046.1 | - | 15 - 16 | 2210 | 83 | ||
AK050226.1 | - | 15 - 16 | 2225 | 83 | ||
AK076066.1 | - | 11 - 12 | 1645 | 83 | ||
AK160937.1 | - | 12 - 13 | 1762 | 83 | ||
BC024824.1 | - | 12 - 13 | 1746 | 5355748 | 83 | |
BC024856.1 | - | 8 - 9 | 866 | 83 | ||
BC026610.1 | - | 9 - 10 | 1316 | 83 | ||
BC034760.1 | - | 15 - 16 | 2177 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |