Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Triml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160412.1 | NP_001153884.1 | 5 - 6 | 679 | 68 | |
GenBank mRNA | AV448075.1 | - | 3 - 4 | 182 | 68 | |
EU599038.1 | - | 7 - 8 | 769 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |