Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtap27-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163105.1 | NP_001156577.1 | 1 | 551 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon |