Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itpripl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163527.1 | NP_001156999.1 | 1 - 2 | 241 | 98 | |
GenBank mRNA | AK028913.1 | - | 1 - 2 | 228 | 98 | |
BC130218.1 | - | 1 - 2 | 90 | 40141227 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |