Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2099940 |
NM_001164201.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2099940 |
DQ358087.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2099940 |
DQ646881.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2325663 |
NM_001164201.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2325663 |
AV270649.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2325663 |
AI429073.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
777344 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
454684 |
AK079397.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
454684 |
AK081206.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
454684 |
AK169810.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
454684 |
BC012512.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
454684 |
NM_175411.4 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
894079 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
574190 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
340302 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
258632 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
648433 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
47864 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
402837 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
930005 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
807313 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
123446 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
58264 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
21375 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
940644 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
282081 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
9372 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
902751 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
68909 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
571356 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
326570 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
877536 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
433131 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
174276 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
438789 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
780798 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
697230 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
876993 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
681591 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
7957 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
573414 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
469180 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
486853 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
285920 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
516361 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
302729 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
651146 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
174281 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
798731 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
472592 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
686575 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
836753 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
411369 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
919734 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
941978 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
811112 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
374584 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
158339 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
465534 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
29283 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
339368 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
674597 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
473807 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
134838 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
103545 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
4668 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
142762 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
35293 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
170032 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
80861 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
147000 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
102140 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
59755 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2099940 |
NM_001164201.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2099940 |
DQ358087.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2099940 |
DQ646881.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2325663 |
NM_001164201.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2325663 |
AV270649.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2325663 |
AI429073.1 |
mouse_transcriptome |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190426 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06078961 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25189972 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190201 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190351 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190110 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190394 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190002 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190191 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06057648 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06642973 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25189977 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190390 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25189973 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190252 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190159 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190117 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190411 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190122 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190331 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190154 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190031 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190415 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190152 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190320 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190410 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190120 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190020 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25189959 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06622158 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190109 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190196 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190357 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06331286 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25189988 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25189969 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25189981 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190119 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190205 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190389 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190398 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06642978 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25189960 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190111 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190324 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190249 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06019469 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190356 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25189970 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190350 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25189995 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190408 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190009 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190425 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25189982 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190400 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190247 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190124 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25189971 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190401 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190392 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25189967 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06078949 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190156 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190195 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190457 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25189991 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190023 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06057667 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190197 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190245 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190250 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190395 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06331296 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06334989 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190322 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190004 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190005 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25189983 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190007 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190243 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190017 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190153 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190033 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190347 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25189990 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190403 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190353 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06573272 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190417 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25189962 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190361 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190189 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06420702 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25189999 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190003 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190029 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05985015 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190346 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190256 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190348 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190328 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06331302 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25189978 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190147 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190151 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190008 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05936474 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190203 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190032 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06420703 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190317 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190422 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25189987 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190254 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190316 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190000 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190387 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190123 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25189964 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190125 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06078952 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190388 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190028 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25189985 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06622157 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190420 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05949514 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190026 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190352 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190349 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190193 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190354 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190253 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190413 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190113 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190360 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190013 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190115 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06420700 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190327 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25190149 |
- |
- |