Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psg18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163685.1 | NP_001157157.1 | 3 | 1654 | 113 | |
GenBank mRNA | AK167661.1 | - | 3 | 1605 | 113 | |
BQ031876.1 | - | 1 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon |