Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.1 ST Array |
10560783 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10560752 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10550770 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10550782 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10560787 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10550782 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10550760 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10560752 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10550770 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10550760 |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0700022266.mm.1 |
TC0700002537.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700022266.mm.1 |
NM_001166757.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022266.mm.1 |
NM_001166757.1 |
Refseq |