Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skint5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167876.1 | NP_001161348.1 | 48 - 49 | 3551 | 79 | |
NM_001167878.1 | NP_001161350.1 | 48 - 49 | 3551 | 79 | ||
GenBank mRNA | EU099301.1 | - | 48 - 49 | 3551 | 79 | |
EU099302.1 | - | 48 - 49 | 3551 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |