Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166376.1 | NP_001159848.1 | 3 | 655 | 78 | |
GenBank mRNA | BC062247.1 | - | 3 | 598 | 1550331 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |