Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700003851.mm.1 |
NM_001025427.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003851.mm.1 |
NM_001025427.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003851.mm.1 |
NM_001039356.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003851.mm.1 |
NM_001039356.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003851.mm.1 |
NM_001166535.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003851.mm.1 |
NM_001166535.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003851.mm.1 |
NM_001166536.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003851.mm.1 |
NM_001166536.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003851.mm.1 |
NM_001166539.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003851.mm.1 |
NM_001166539.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003851.mm.1 |
NM_001166540.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003851.mm.1 |
NM_001166540.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003851.mm.1 |
NM_001166541.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003851.mm.1 |
NM_001166541.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003851.mm.1 |
NM_001166542.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003851.mm.1 |
NM_001166542.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003851.mm.1 |
NM_001166543.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003851.mm.1 |
NM_001166543.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003851.mm.1 |
NM_001166544.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003851.mm.1 |
NM_001166544.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003851.mm.1 |
NM_001166545.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003851.mm.1 |
NM_001166545.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003851.mm.1 |
NM_001166546.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003851.mm.1 |
NM_001166546.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003851.mm.1 |
TC1700000461.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700003864.mm.1 |
NM_001039356.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003864.mm.1 |
NM_001039356.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003864.mm.1 |
NM_001166537.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003864.mm.1 |
NM_001166537.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003864.mm.1 |
NM_001166539.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003864.mm.1 |
NM_001166539.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003864.mm.1 |
NM_016660.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003864.mm.1 |
NM_016660.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003864.mm.1 |
TC1700000461.mm.1 |
MTA TC |