Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.1 ST Array |
10560395 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10560397 |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0700021638.mm.1 |
TC0700002458.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021638.mm.1 |
NM_054058.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021638.mm.1 |
NM_054058.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021645.mm.1 |
TC0700002458.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021645.mm.1 |
NM_054058.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021645.mm.1 |
NM_054058.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021637.mm.1 |
TC0700002458.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021637.mm.1 |
NM_054058.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021637.mm.1 |
NM_054058.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021647.mm.1 |
TC0700002458.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021647.mm.1 |
NM_054058.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021647.mm.1 |
NM_054058.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021646.mm.1 |
TC0700002458.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021646.mm.1 |
NM_054058.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021646.mm.1 |
NM_054058.1 |
Refseq |