Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182694.2 | NP_874353.2 | 2 - 3 | 2475 | 79 | |
NM_182696.2 | NP_874355.1 | 2 - 3 | 431 | 79 | ||
GenBank mRNA | BC089358.1 | - | 2 - 3 | 2475 | 30610755 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |